Learn More
Biolegend Spark BlueTM 515 anti-human CD
Supplier:  Biolegend 344137
Encompass_Preferred
Spark Blue™ 515 anti-human CD141 (Thrombomodulin) [M80]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-274-5677