Learn More
Biolegend Spark RedTM 718 anti-human CD1
Supplier:  Biolegend 301875
Encompass_Preferred
Spark Red™ 718 anti-human CD14 [M5E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-290-6337