Learn More
Biolegend Spark UVTM 387 anti-human CD16
Supplier:  Biolegend 302077
Encompass_Preferred
Spark UV™ 387 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-272-2502