Learn More
Medchemexpress LLC HY-17621 5mg Medchemexpress, Sparsentan CAS:254740-64-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176215MG
Encompass_Preferred
Medchemexpress, HY-17621 5mg Sparsentan CAS:254740-64-2 Sparsentan (RE-021) is a highly potent dual angiotensin II and endothelin A receptor antagonist with Kis of 0.8 and 9.3 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2002