Learn More
Chemglass Life Sciences Spatula, Stainless Steel #304, 7" OAL, Non-Magnetic, Tapered End
SDP
Supplier:  Chemglass Life Sciences CG198413
Encompass_Preferred
Spatula will pass through a 14/10 or 14/20 outer joint and is made of non-magnetic stainless steel having a mirror finish with an overall length of 7in (178mm). One end is rounded, the other is tapered to a point approximately 3/32in (2.5mm). Both ends are approximately 5/16in (8mm) wide x 2in (51mm) in length.
Catalog No. 50-873-390