Learn More
Medchemexpress LLC SPD304 dihydrochloride | 1049741-03-8 | MFCD09265259 | 99.8% | 620.53 | C32H34Cl2F3N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY111255A5MG
Encompass_Preferred
SPD304 dihydrochloride is a small-molecule selective tumor necrosis factor-alpha (TNF-α) inhibitor that promotes dissociation of TNF trimers and blocks interaction with TNFR1. It is supplied as a high-purity research reagent intended for in vitro biochemical and cell-based studies of TNF signaling and inhibitor effects.
  • Selective TNF-α inhibition by promoting TNF trimer dissociation.
  • Characterized activity with reported IC50 of 22 μM for TNFR1 binding inhibition.
  • High purity suitable for in vitro biochemical and cell-based assays.
  • Available in small milligram quantities for research use.
  • Useful as a tool compound to study TNF signaling and receptor interactions.
Catalog No. 50-002-63607