Learn More
Medchemexpress LLC Spdb | 115088-06-7 | MFCD20229028 | 98.0% | 326.40 g·mol⁻¹ | C13H14N2O4S2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY12448200MG
Encompass_Preferred
SPDB is a glutathione-cleavable antibody-drug conjugate (ADC) linker provided for research use. It is designed to remain stable in circulation and be cleaved intracellularly by glutathione to release payloads, making it suitable for ADC synthesis, conjugation method development, and linker-cleavage studies.
  • Glutathione-cleavable linker
  • Stable in circulation, labile in target cells
  • Suitable for ADC synthesis and linker-cleavage studies
  • High purity (~98.0%) for reproducible results
  • Available in multiple package sizes, including 200 mg
  • Store sealed, away from moisture; recommended -20°C
Catalog No. 50-002-03751