Learn More
Medchemexpress LLC Spdmv | 890409-85-5 | 96.4% | 354.44 g/mol | C15H18N2O4S2 | 100 MG

Supplier: Medchemexpress LLC HY129368100MG
SPDMV is a glutathione-cleavable antibody-drug conjugate (ADC) linker supplied for research use. It is designed to connect antibodies to cytotoxic payloads and release the payload intracellularly via glutathione-mediated cleavage.
- Glutathione-cleavable disulfide linker.
- Intended for ADC synthesis and conjugation chemistry.
- Purity 96.37%.
- CAS number 890409-85-5.
- Molecular weight 354.44 g/mol.
- Chemical formula C15H18N2O4S2.
- Available in 100 MG vial.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.