Learn More
Sorbent Technologies Inc SPE Cartridges, Chromabond LV, Alumina N, 15ml, 1000mg, 30/pk
SDP
Supplier:  Sorbent Technologies Inc 732091
Encompass
SPE Cartridges, Chromabond LV, Alumina N - Applications: Together with a strongly acidic cation exchanger for PCB and pesticides. Large volume PP columns with PE filter elements. Funnel-shaped reservoir with 15 ml volume. Especially for clinical samples-the whole sample (e. g. urine, serum, blood) can be applied to the column in one step. Can be directly used in the Zymate lab robots of Zymark.
Catalog No. NC1187663