Learn More
Cayman Chemical SPE Cartridges (C-18) (6 ml), 25 ea
SDP
Supplier:  Cayman Chemical 40002025
Encompass_Preferred
This product may be used for purification of various lipid mediators such as prostaglandins and hydroxylated fatty acids from various sample types prior to analysis by LC-MS/MS or ELISA. References are provided below for applicable C-18 sample purification methods prior to LC-MS analysis.{19261,26615,8825,15162} For sample purification prior to analysis by ELISA, we recommend consulting the appropriate assay kit booklet for instructions.
Catalog No. NC9057503