Learn More
Sigma Aldrich Fine Chemicals Biosciences Spe I from Sphaerotilus sp
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11008951001
Encompass
Spe I recognizes the sequence *A8595*CTAGT and generates fragments with 5-cohesive termini.
Catalog No. 50-290-5466