Learn More
Essendant Spracht MIC2010 Digital USB Microphone, Black
Supplier:  Essendant SPTMIC2010
Encompass_Preferred
Perfect Huddle room microphone Set the mic on the table plug into the USB port of your PC/Mac and your microphone is ready for Skype teams Zoom GoToMeeting WebEx other VoIP calls Record audio for YouTube videos or podcasts too Why use an external microphone instead of your computer s Well everyone will be heard better since the microphone can be ideally located between all the people in the room USB mics process sound internally they act as their own sound card when attached to a computer A USB mic will produce the same quality output regardless of the computer s hardware specs they ll work better than analog mics on computers with an inexpensive sound card or none at all Cord Length- 6 ft Power Source(s)- USB Cable Batteries Required- No Operating System Compatibility- Mac OS X Windows XP Vista 7 8 10
Catalog No. 50-000-27975