Learn More
Medchemexpress LLC SPEB E.coli His-SU 5ug

Supplier: Medchemexpress LLC HYP720725UG
Recombinant SPEB protein with an N-terminal His-SUMO tag, expressed in Escherichia coli and supplied lyophilized for research use. The manufacturer datasheet provides the full amino acid sequence, formulation details, and recommended handling and storage instructions to support biochemical and enzymatic studies.
- Recombinant protein expressed in Escherichia coli.
- N-terminal His-SUMO tag facilitates affinity purification.
- Approximately 49.5 kDa apparent molecular weight.
- Lyophilized from PBS with 6% trehalose for improved stability.
- Recommended storage at -20°C for long-term; aliquot and store at -80°C for extended storage.
- Full amino acid sequence and handling instructions provided in the datasheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.