Learn More
Bachem H-Ala-AMC
SDP
Supplier:  Bachem 4028736.0050
Encompass_Preferred
H-Ala-AMC, 77471-41-1, C13H14N2O3, Mr 246.27, 50mg. H-Ala-MCA (free base). Special fluorogenic substrate for aminopeptidase M (microsomal alanyl aminopeptidase, membrane aminopeptidase, APN). Furthermore, the major aminopeptidase from human skeletal muscle showed highest activity with this substrate.
Catalog No. 50-194-7001