Learn More
Perkin Elmer US LLC Special Sample Pan Manipulator - Suction
Supplier:  Perkin Elmer US LLC B0142512
Encompass_Preferred
Sample Pan Manipulators are specially designed to simplify picking up and positioning sample pans and covers. Their use is essential to preventing sample contamination.
Catalog No. 50-210-4150