Learn More
Medicus Health Specimen Transport Hardside Cooler, 16 Quart, White
SDP
Supplier:  Medicus Health 2934M2WHITE
Encompass
• Hardside, 16-quart, non-labeled specimen transport medical cooler is perfect for transporting specimens to and from the lab while maintaining temperatures
• Small size makes it light weight and easy to transport
Specimen Transport Hardside Cooler is White • Insulated cooler offers superior protection for your specimens or blood products
• Leak resistant sturdy latches and hinges
• Durable gasket locks in the cold to preserve specimens during transport to and from the lab
• Smooth non-absorbent sides makes it easy to clean and disinfect
• Convenient handle on top can be used to secure lid during transport
• Cooler can also be used to transport temperature sensitive medications without leaking
• Great option for Blood Banks to send units to the ER or surgery to help prevent units from returning over temperature
• BPA Free and Lifetime Warranty
• External Dimensions: 16.7"W x 12.8"D x 11.2"H; Internal Dimensions: 13"W x 9"D x 9"H
Catalog No. NC3898564