Learn More
Electron Microscopy Sciences Histo-Tote™ Specimen Transport System
SDP
Supplier:  Electron Microscopy Sciences 64229
Encompass_Preferred
Acrylic, modular tray for specimen transport. Facilitates safe and efficient transport of prefilled specimen containers. Integral handles allow for easy and safe transport. Accomodates 20 ml, 40 ml, and 60 ml prefilled specimen containers. Two inserts per tray.
Catalog No. 50-192-8619