Learn More
Medchemexpress LLC Specneuzhenide | 449733-84-0 | 20 MG

Supplier: Medchemexpress LLC HYN607520MG
Specneuzhenide (Nuezhenide) is a high-purity phenol glycoside (99.87%) isolated from Ligustrum sinense. This white to off-white solid possesses anti-tumor activity, specifically against human HepG2 cells, with an IC50 > 100 μM. Its molecular weight is 686.65 with the chemical formula C31H42O17.
- Isolated from Ligustrum sinense
- Exhibits anti-tumor activity
- High purity of 99.87%
- White to off-white solid appearance
- Soluble in DMSO at ≥ 100 mg/mL
- Store at 4°C protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.