Learn More
TARGETMOL CHEMICALS INC SPECNUEZHENIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T38035MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Specnuezhenide (Nuezhenide) significantly protects human neuroblastoma SH-SY5Y cells from 6-hydroxydopamine-induced neurotoxicity.Nuezhenide is the main inhibitory compound of fruits it shows a clear inhibitory effect to the seeds of wheat and Chinese cabbage. purity: 99%
Catalog No. 50-257-6872