Learn More
Alkali Scientific Spectinomycin Dihydrochloride Pentahydrate, 5 G
Supplier: Alkali Scientific SPE005CP
Spectinomycin Dihydrochloride Pentahydrate, 5g, is a highly concentrated antibacterial agent available in powder form for laboratory and clinical applications. It is primarily used to treat infections caused by Gram-negative bacteria, most notably in the treatment of Neisseria gonorrhoeae. Spectinomycin is often used when other antibiotics are ineffective, particularly in penicillin-resistant strains. The 5g size is perfect for small-scale research experiments in microbiology and molecular biology. It is commonly used in bacterial culture media to inhibit protein synthesis and is an important tool in studying bacterial growth and resistance mechanisms.