Learn More
Quanterix Inc SPECTRAL DAPI

Supplier: Quanterix Inc FP1490
- Nuclear counterstain for clear visualization of cell nuclei.
- Optimized for seamless integration within Multiplex IHC workflows.
- Enables simultaneous detection of up to 6 or 8 tissue biomarkers.
- Compatible with standard IHC laboratory procedures.
- Works with formalin-fixed paraffin-embedded (FFPE) tissue.
- Validated for use with multispectral imaging systems.
- Unit size of 1.5 mL.
- Shipping condition is Blue Ice.
- Compatible with human and mouse tissue samples.
- Excitation wavelength of 368 nm and emission wavelength of 461 nm.
- Automation compatible.
- Product category: Reagent Pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.