Learn More
Anton Paar Cora 5001 direct raman spectrometer with sample compartment, wavelengths 532/1064nm
Supplier: Anton Paar 225552
Single Wavelength or Dual Wavelength.Available wavelengths:532nm785nm1064nmBattery operation option