Learn More
Anton Paar Cora 5001 direct raman spectrometer with sample compartment, wavelength 1064nm
Supplier: Anton Paar 225604
Single Wavelength or Dual Wavelength.Available wavelengths:532nm785nm1064nmBattery operation option