Learn More
Med Vet International Dynarex Vaginal Specula Disposable w/ Light Option, Large, 80/Case
Supplier:  Med Vet International 4916
Encompass_Preferred
Dynarex Vaginal Speculums are constructed from high-strength plastic for optimal reliability and feature a smooth, rounded design for improved patient comfort. The Speculums are designed to help facilitate safe and effective visualization during pelvic exams. The Speculums are easy to adjust and hold in a variety of positions for convenient one-hand operation. Dynarex Vaginal Speculums are single-use and disposable for optimal hygiene. Dynarex Vaginal Speculums are premium-quality disposable speculums designed to help provide safe, effective visualization during pelvic exams. These smooth, rounded Speculums are designed to adjust to a variety of positions for optimal convenience. 
Catalog No. 50-282-9721
 Restricted Item