Learn More
Grainger Speed Control 115V AC 3 A Dial
Supplier:  Grainger 14U187
Encompass_Preferred
The Grainger Speed Control is a 115V AC dial speed controller with a maximum load rating of 3 amps. It allows users to adjust the speed of single phase AC motors and other electrical loads by varying the voltage with a dial knob. The enclosure is made of plastic with screw terminal connections. Key features include overload and short circuit protection. It can be mounted on walls or panels. This provides variable speed control for applications like fans, pumps, conveyors, and other AC motor-driven equipment.
Catalog No. 50-275-1718