Learn More
Grainger Speed Control 125V AC 3 A Dial
Supplier:  Grainger 14U188
Encompass_Preferred
The Grainger Approved Speed Control is a single-phase AC motor speed controller that provides adjustable speed for fractional horsepower fan and blower motors. It has a maximum load rating of 1 HP at 120V AC and a speed range of 50-100%. This electronic speed control features a rotary knob for easy speed adjustment and has thermal overload protection. It measures 4-1/2" L x 2-3/4" W x 2-1/4" H and has a 6 foot power cord with 3-prong plug. The speed control requires a minimum load of 40 watts and can handle up to 8.3 Amps. It is cULus listed and ideal for controlling the speed of fans, blowers, pumps and other AC motors in both commercial and residential applications.
Catalog No. 50-275-3315