Learn More
Biotage SPEFRT PEFRITS 10U/70ML 100
SDP
Supplier:  Biotage 1201067F
Encompass
Frit; Biotage; Isolute; For SPE; Polyethylene; 10um; 27mm (1.062 in.) dia.; Column type F, 70mL; 100/pack
Catalog No. 50-231-1377