Learn More
FUJIFILM IRVINE SCIENTIFIC INC SPERM WASHING MEDIUM PK/12ML
Supplier: FUJIFILM IRVINE SCIENTIFIC INC 998312X12ML
Sperm Washing Medium supports all steps of sperm preparation prior to fertilization, whether it is IUI, IVF, or other ART procedures. It is designed to maintain sperm quality on the tabletop without CO2 incubation. Sperm Washing Medium provides lot-to-lot consistency for sperm processing through its quality formulation, which is stringently tested by mouse embryo assay (MEA). 12x12ML.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.