Learn More
Cayman Chemical SpermIdIn-d6 5mg
SDP
Supplier:  Cayman Chemical 346975 MG
Encompass_Preferred
An internal standard for the quantification of spermidine by GC- or LC-MS
Catalog No. 50-291-4049