Learn More
Apexbio Technology LLC Spermidine 124-20-9 1mL(10 mM in Water)
SDP
Supplier:  Apexbio Technology LLC BA74085.1
Encompass_Preferred
Spermidine (CAS 124-20-9) is a naturally occurring polyamine that contributes to cellular homeostasis It stabilizes cell membranes and enhances the activity of antioxidant enzymes thereby supporting cellular defense mechanisms against oxidative stress Spermidine has been shown to improve the function and gene expression of photosystem II (PSII) while reducing the intracellular levels of reactive oxygen species such as hydroxyl radicals and superoxide anions Due to these properties spermidine is widely utilized in biomedical research examining oxidative stress regulation cellular aging and related physiological processes
Catalog No. 50-000-87867