Learn More
Medchemexpress LLC Spermidine-13C4 hydrochloride | 1313734-84-7 | 95% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1776AS11MG
Encompass_Preferred
Spermidine-13C4 (hydrochloride) is the 13C-labeled Spermidine trihydrochloride. Spermidine hydrochloride maintains cell membrane stability, increases antioxidant enzyme activities, improving photosystem II (PSII), and relevant gene expression. It significantly decreases H2O2 and O2.- contents.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12803