Learn More
Medchemexpress LLC Spermidine-d6 | 2514812-10-1 | 96.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1776S5MG
Encompass_Preferred
Spermidine-d6 is the deuterium labeled Spermidine, designed for research applications. It plays a role in maintaining cell membrane stability, increasing antioxidant enzyme activities, and improving photosystem II (PSII) and relevant gene expression. This compound significantly decreases H2O2 and O2.- contents. Deuteration, incorporating stable heavy isotopes into drug molecules, has gained attention for its potential to affect pharmacokinetic and metabolic profiles of drugs, making Spermidine-d6 valuable as a tracer and internal standard.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Maintains cell membrane stability
  • Increases antioxidant enzyme activities
  • Improves photosystem II (PSII) and relevant gene expression
  • Decreases H2O2 and O2.- contents
Catalog No. 50-004-11716