Learn More
Medchemexpress LLC Spermine(N3BBB) | 1190203-80-5 | C25H48N6O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151657
Encompass_Preferred
Spermine(N3BBB) is a click chemistry reagent containing an azide group. It is utilized for various biochemical studies, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Suitable for various biochemical studies
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Can be used as an ADC linker
Catalog No. 50-003-74396