Learn More
Medchemexpress LLC Spermine(N3BBB) | 1190203-80-5 | C25H48N6O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516575MG
Encompass_Preferred
Spermine(N3BBB) is a click chemistry reagent containing an azide group and can be used for various biochemical studies.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups.
  • Participates in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
Catalog No. 50-003-72173