Learn More
Apexbio Technology LLC Spermine tetrahydrochloride 306-67-2 100mg
SDP
Supplier:  Apexbio Technology LLC B6522100
Encompass_Preferred
Spermine tetrahydrochloride (CAS 306-67-2) is an endogenous polyamine involved in cellular metabolism and homeostasis It modulates enzyme activities and signaling pathways particularly influencing cell cycle progression and nucleic acid stability In biomedical research Spermine tetrahydrochloride is commonly employed to investigate mechanisms regulating cell proliferation DNA and RNA interactions and the cellular response in oncological models Its ability to interfere with these processes makes it a valuable reagent for studies on cancer biology and the regulation of cell growth
Catalog No. 50-000-89243