Learn More
Waters Corp Spherisorb Cyano (CN) Column, Normal Phase, 80 à , 5 µm, 4.6 mm X 250 mm, 1/pk, with eConnect Column Tag
Supplier:  Waters Corp PSS830915RF
Encompass_Preferred
Spherisorb Cyano (CN) Column, Normal Phase, 80 à , 5 µm, 4.6 mm X 250 mm, 1/pk, with eConnect Column Tag
Catalog No. 50-239-1307