Learn More
Waters Corp Spherisorb Cyano (CN) OBD Prep Column, 80Å, 5 μm, 10 mm x 250 mm
Supplier:  Waters Corp 186008287
Encompass_Preferred
This column features Cyano (CN) chemistry with a propyl cyano bonded ligand to a silica substrate. It is conditioned with normal phase solvent and designed for demanding HPLC assays, offering a wide pH range for various analytes. The OBD prep column design provides direct scalability for method transfer.
  • Features propyl cyano bonded ligand to a silica substrate.
  • Conditioned and shipped with normal phase solvent.
  • Handles demanding chromatographic conditions.
  • Offers a wide pH range (2 pH to 8 pH).
  • Enables direct scalability for method transfer with OBD prep design.
  • Maintains chemical and mechanical stability under demanding conditions.
  • Cited in over 2,000 analytical abstracts.
Catalog No. 50-239-1027