Learn More
Waters Corp Waters Column, Cyano (CN), Reversed Phase, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp PSS830909
Encompass_Preferred
The Spherisorb CN chemistry features a general-purpose reversed-phase propyl cyano bonded ligand on a silica substrate. This column is conditioned and shipped with acetonitrile, designed to provide excellent peak symmetry and efficiency, outstanding reproducibility, and an enhanced column lifetime.
  • Useful when highly hydrophobic compounds cannot be eluted with standard C18 and C8 columns or typical reversed-phase eluents.
  • Provides rapid elution for hydrophobic molecules.
  • Offers unique selectivity for polar compounds.
  • Delivers excellent peak shape for strongly basic compounds.
  • Exhibits different selectivity compared to normal C18 and C8 reversed-phase columns.
  • Manufactured under cGMP.
  • ISO 9002 certified.
  • Registered with the FDA as a manufacturer of Class 1 medical devices.
Catalog No. 50-819-133