Learn More
Medchemexpress LLC Sphingosine-1-phosphate | 26993-30-6 | 99.5% | 379.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY108496S500UG
Encompass_Preferred
Sphingosine-1-phosphate (S1P) acts as an agonist for S1P1-5 receptors and a ligand for GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. This lipid mediator stimulates DNA synthesis, cell proliferation, and migration.
  • Agonist for S1P1-5 receptors and ligand for GPR3, GPR6, GPR12.
  • Functions as an intracellular second messenger.
  • Mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors.
  • Stimulates DNA synthesis, cell proliferation, and migration.
  • Derived from sphingomyelin or other membrane phospholipids.
Catalog No. 50-003-46889