Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sphingosine-1-phosphate (d17:1) | 474923-27-8 | 99.0% | C17H36NO5P | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1416301MG

Encompass_Preferred

Sphingosine-1-phosphate d17:1 is a ceramide derivative primarily used in cancer research.

  • Derivative of ceramide
  • Suitable for cancer research
  • Purity of 99.0%
  • Molecular weight of 365.45
  • Chemical formula: C17H36NO5P
  • Appears as a white to off-white solid
  • Soluble in 0.3 M NaOH (4 mg/mL)
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-56513


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.