Learn More
Medchemexpress LLC Sphingosine-1-phosphate (S1P) | 26993-30-6 | 99.0% | 379.47 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10849610MG
Encompass_Preferred
Sphingosine-1-phosphate (S1P) is an agonist of S1P1-5 receptors and a ligand of GPR3, GPR6, and GPR12. It functions as an intracellular second messenger and mobilizes Ca2+ as an extracellular ligand for G protein-coupled receptors. S1P is a significant lipid mediator produced from Sphingomyelin or other membrane phospholipids.
  • Stimulates DNA synthesis
  • Promotes cell proliferation
  • Facilitates cell migration
Catalog No. 50-003-46731