Learn More
Selleck Chemical LLC Sphingosine S2210-1g
SDP
Supplier:  Selleck Chemical LLC S22101G
Encompass_Preferred
Sphingosine (2-amino-4-trans-octadecene-1 3-diol C18 Sphingosine) is an 18-carbon amino alcohol with an unsaturated hydrocarbon chain which forms a primary part of sphingolipids a class of cell membrane lipids that include sphingomyelin an important phospholipid
Catalog No. 50-311-2786