Learn More
Cayman Chemical Sphingosine d17 1
SDP
Supplier:  Cayman Chemical 1000790210 MG
Encompass_Preferred
A naturally-occurring but uncommon form of sphingosine, accounting for approximately 13% of the sphingosine in human skin; can be phosphorylated by sphingosine kinases to produce C17 sphingosine-1-phosphate
Catalog No. 50-288-9014