Learn More
Cayman Chemical Sphingosine Kinase 1 Blocking
SDP
Supplier:  Cayman Chemical 100068231 EA
Encompass_Preferred
Peptide Sequence: human SPHK1 amino acids 264-274 (DLESEKYRRLG) · To be used in conjunction with Cayman’s Sphingosine Kinase 1 polyclonal antibody (Item No. 10006822) to block protein-antibody complex formation during immunochemical analysis of SPHK1 protein.
Catalog No. 50-288-6758