Learn More
Cayman Chemical Sphingosine Kinase 1 human
SDP
Supplier:  Cayman Chemical 1034825 UG
Encompass_Preferred
Source: Human recombinant N-terminal His-tagged protein from Sf9 cells • SPHK1 catalyzes the production of sphingosine-1-phosphate, a lipid mediator with broad spectrum of biological activities including cell proliferation, survival, migration, cytoskeletal organization, and morphogenesis.
Catalog No. 50-289-0513