Learn More
Cayman Chemical Sphingosine Kinase Inhibitor
SDP
Supplier:  Cayman Chemical 1000922225 MG
Encompass_Preferred
An SPHK1 inhibitor (IC50 = 0.5 µM for non-ATP-competitive inhibition of human recombinant SPHK1); selective for SPHK1 over ERK2, PI3K, and PKCα at 50s = 0.9-4.6 µM)
Catalog No. 50-289-0909