Learn More
Cayman Chemical Sphingosine1phosphated7 d18
SDP
Supplier:  Cayman Chemical 100081211 MG
Encompass_Preferred
An internal standard for the quantification of S1P by GC- and LC-MS
Catalog No. 50-288-8620