Learn More
Medchemexpress LLC Sphingosine kinase 1 (SPHK1) protein, human (Sf9, 363 a.a.) | >90.0% | 10UG

Supplier: Medchemexpress LLC HYP70276510UG
Recombinant human sphingosine kinase 1 (SPHK1), 363 amino acids, produced in Sf9 insect cells. Supplied as a tag-free, 0.22 μm filtered solution in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT. Intended for biochemical assays and enzymology; shipped on dry ice and recommended for storage at -80 °C.
- Recombinant human protein expressed in Sf9 insect cells.
- 363 amino acids, approximately 40.3 kDa molecular weight.
- Tag-free formulation suitable for functional and binding assays.
- Purity >90.0% as determined by reducing SDS-PAGE.
- Supplied in stabilized buffer with glycerol and DTT to preserve activity.
- Shipped on dry ice; store at -80 °C and avoid repeated freeze-thaw cycles.
- Available in multiple size options to suit assay scale.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.