Learn More
Cayman Chemical SphngolIpId MIxturebovIn 1mL
SDP
Supplier:  Cayman Chemical 293751 ML
Encompass_Preferred
A mixture of isolated bovine neuronal sphingolipids that contains hydroxy- and non-hydroxy galactosylceramides, sulfatides, and sphingomyelins; intended for use as a reference standard for the identification of these lipids in various sample types by TLC, LC-, or GC-MS
Catalog No. 50-299-0197