Learn More
Cayman Chemical SphngomyelIn Synthase 2hum
SDP
Supplier:  Cayman Chemical 41070100 UG
Encompass_Preferred
Source: Recombinant N-terminal mouse IgG1 Fc-tagged human SMS2 expressed in HEK293 cells • Amino acids: 1-79 • MW: 35.7 kDa
Catalog No. 50-294-4790